Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abtb1
Entrez Gene ID
80283
Gene Name
ankyrin repeat and BTB (POZ) domain containing 1
Gene Aliases
AI847549, BC003234, BPOZ, EF1ABP
Chromosome Location
Chr.6: 88835914 - 88842386 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030251.3 NP_084527.2 11 - 12 1347 63
GenBank mRNA AB053477.1 - 11 - 12 1278 63
AK049100.1 - 11 - 12 1243 63
AK088437.1 - 11 - 12 1312 63
AK155432.1 - 5 - 6 4161 63
AK155505.1 - 5 - 6 4161 63
AK155644.1 - 5 - 6 4161 63
AK220208.1 - 10 - 11 2583 63
BC003234.1 - 11 - 12 1291 63

View Full Product Details