The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025466.1 | NP_079742.1 | 5 - 6 | 502 | 101 |
GenBank mRNA | AK008622.1 | - | 5 - 6 | 502 | 101 |
AK008640.1 | - | 5 - 6 | 500 | 101 | |
AK008641.1 | - | 5 - 6 | 502 | 101 | |
AK008647.1 | - | 5 - 6 | 499 | 101 | |
AK008722.1 | - | 5 - 6 | 500 | 101 | |
AK008745.1 | - | 5 - 6 | 502 | 101 | |
AK008933.1 | - | 5 - 6 | 504 | 101 | |
AK008956.1 | - | 5 - 6 | 499 | 101 | |
AK008990.1 | - | 5 - 6 | 504 | 101 | |
AK009145.1 | - | 5 - 6 | 500 | 101 | |
AK028136.1 | - | 5 - 6 | 498 | 101 | |
AY456960.1 | - | 5 - 6 | 502 | 101 | |
BC099446.1 | - | 5 - 6 | 485 | 101 | |
BC132540.1 | - | 5 - 6 | 492 | 101 | |
BC132542.1 | - | 5 - 6 | 492 | 101 |