Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tulp3
Entrez Gene ID
22158
Gene Name
tubby-like protein 3
Gene Aliases
2310022L06Rik, AI316887
Chromosome Location
Chr.6: 128321161 - 128355872 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011657.2 NP_035787.1 4 - 5 500 65
GenBank mRNA AB016963.1 - 4 - 5 443 65
AF045582.1 - 4 - 5 453 65
BC060068.1 - 4 - 5 430 65

View Full Product Details