Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BC048546
Entrez Gene ID
232400
Gene Name
cDNA sequence BC048546
Gene Aliases
Ovos
Chromosome Location
Chr.6: 128539822 - 128581606 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001179.3 NP_001001179.2 1 - 2 163 73
GenBank mRNA AK138836.1 - 1 - 2 164 73
AK161184.1 - 1 - 2 166 73

View Full Product Details