The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170485.1 | NP_001163956.1 | 2 - 3 | 191 | 72 |
NM_001170486.1 | NP_001163957.1 | 3 - 4 | 303 | 72 | |
NM_026552.3 | NP_080828.1 | 4 - 5 | 422 | 72 | |
GenBank mRNA | AJ278129.1 | - | 4 - 5 | 348 | 72 |
AK017993.1 | - | 4 - 5 | 423 | 72 | |
AK018069.1 | - | 4 - 5 | 360 | 72 | |
AK039215.1 | - | 4 - 5 | 370 | 72 | |
AK076248.1 | - | 4 - 5 | 370 | 72 | |
AK077325.1 | - | 4 - 5 | 344 | 72 | |
AK133626.1 | - | 4 - 5 | 370 | 72 | |
AK136082.1 | - | 4 - 5 | 365 | 72 | |
AK150690.1 | - | 4 - 5 | 356 | 72 | |
AK153380.1 | - | 4 - 5 | 354 | 72 | |
AK159410.1 | - | 3 - 4 | 251 | 72 | |
AK166440.1 | - | 4 - 5 | 344 | 72 | |
BC015280.1 | - | 3 - 4 | 334 | 72 | |
BC055309.1 | - | 3 - 4 | 333 | 72 |