The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001036684.2 | NP_001031761.1 | 18 - 19 | 3536 | 96 |
NM_009723.5 | NP_033853.1 | 20 - 21 | 3725 | 96 | |
GenBank mRNA | AB030737.1 | - | 17 - 18 | 3097 | 96 |
AB030738.1 | - | 17 - 18 | 3097 | 96 | |
AF053471.1 | - | 19 - 20 | 3554 | 96 | |
AK054112.1 | - | 22 - 23 | 3823 | 96 | |
AK147240.1 | - | 20 - 21 | 3611 | 96 | |
AK147357.1 | - | 18 - 19 | 3465 | 96 | |
AK147369.1 | - | 20 - 21 | 3660 | 96 | |
AK147399.1 | - | 18 - 19 | 3688 | 96 | |
AK147501.1 | - | 20 - 21 | 3672 | 96 | |
GU734816.2 | - | 20 - 21 | 3232 | 96 |