The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170673.3 | NP_733773.2 | 1 - 2 | 233 | 71 |
GenBank mRNA | AK042692.1 | - | 1 - 2 | 234 | 71 |
AK047571.1 | - | 1 - 2 | 177 | 71 | |
AK137422.1 | - | 1 - 2 | 234 | 71 | |
BB639866.1 | - | 1 - 2 | 234 | 71 | |
BC115728.1 | - | 1 - 2 | 183 | 71 |