The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172161.4 | NP_751893.3 | 3 - 4 | 507 | 117 |
GenBank mRNA | AJ440756.2 | - | 3 - 4 | 426 | 117 |
AK028733.1 | - | 3 - 4 | 471 | 117 | |
AK150056.1 | - | 3 - 4 | 447 | 117 | |
AK154715.1 | - | 3 - 4 | 355 | 117 | |
AY162378.1 | - | 3 - 4 | 486 | 117 | |
AY162381.1 | - | 2 - 3 | 303 | 117 | |
BC085324.1 | - | 3 - 4 | 332 | 117 |