The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027526.1 | NP_081802.1 | 12 - 13 | 1505 | 62 |
GenBank mRNA | AK018120.1 | - | 11 - 12 | 1336 | 62 |
AK158756.1 | - | 13 - 14 | 1531 | 62 | |
BC138284.1 | - | 12 - 13 | 1471 | 62 | |
BC145232.1 | - | 12 - 13 | 1498 | 62 | |
BC145752.1 | - | 12 - 13 | 1471 | 62 |