Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bms1
Entrez Gene ID
213895
Gene Name
BMS1 homolog, ribosome assembly protein (yeast)
Gene Aliases
AA408648, AU020092, AU043373, BB007109, BC030906, Bms1l, mKIAA0187
Chromosome Location
Chr.6: 118383381 - 118420018 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194339.1 NP_919320.1 10 - 11 2155 79
GenBank mRNA AK129082.1 - 10 - 11 2115 79
AK133013.1 - 10 - 11 2105 79
AK134315.1 - 10 - 11 2163 79
AK135031.1 - 9 - 10 2652 79
AK162178.1 - 10 - 11 2213 79
BC030906.1 - 3 - 4 955 79
BC057054.1 - 10 - 11 2155 79

View Full Product Details