Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bms1
Entrez Gene ID
213895
Gene Name
BMS1 homolog, ribosome assembly protein (yeast)
Gene Aliases
AA408648, AU020092, AU043373, BB007109, BC030906, Bms1l, mKIAA0187
Chromosome Location
Chr.6: 118383381 - 118420018 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194339.1 NP_919320.1 17 - 18 3128 88
GenBank mRNA AK129082.1 - 17 - 18 3088 88
AK133013.1 - 17 - 18 3078 88
AK162178.1 - 17 - 18 3186 88
BC030906.1 - 10 - 11 1928 88
BC057054.1 - 17 - 18 3128 88

View Full Product Details