Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bms1
Entrez Gene ID
213895
Gene Name
BMS1 homolog, ribosome assembly protein (yeast)
Gene Aliases
AA408648, AU020092, AU043373, BB007109, BC030906, Bms1l, mKIAA0187
Chromosome Location
Chr.6: 118383381 - 118420018 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194339.1 NP_919320.1 3 - 4 525 108
GenBank mRNA AK028266.1 - 3 - 4 524 108
AK129082.1 - 3 - 4 485 108
AK133013.1 - 3 - 4 475 108
AK134315.1 - 3 - 4 533 108
AK135031.1 - 3 - 4 513 108
AK162178.1 - 3 - 4 583 108
BC057054.1 - 3 - 4 525 108

View Full Product Details