Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bms1
Entrez Gene ID
213895
Gene Name
BMS1 homolog, ribosome assembly protein (yeast)
Gene Aliases
AA408648, AU020092, AU043373, BB007109, BC030906, Bms1l, mKIAA0187
Chromosome Location
Chr.6: 118383381 - 118420018 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194339.1 NP_919320.1 8 - 9 1259 63
GenBank mRNA AK028266.1 - 8 - 9 1258 63
AK129082.1 - 8 - 9 1219 63
AK133013.1 - 8 - 9 1209 63
AK134315.1 - 8 - 9 1267 63
AK135031.1 - 7 - 8 1756 63
AK162178.1 - 8 - 9 1317 63
BC030906.1 - 1 - 2 59 63
BC057054.1 - 8 - 9 1259 63

View Full Product Details