Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
M1ap
Entrez Gene ID
110958
Gene Name
meiosis 1 associated protein
Gene Aliases
D6Mm5e
Chromosome Location
Chr.6: 82946908 - 83030309 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033079.2 NP_149070.2 9 - 10 1515 76
GenBank mRNA AF084364.1 - 8 - 9 1484 76
AK131954.1 - 9 - 10 1515 76
BC119803.1 - 8 - 9 1442 76
BC122521.1 - 8 - 9 1447 76

View Full Product Details