Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dqx1
Entrez Gene ID
93838
Gene Name
DEAQ RNA-dependent ATPase
Gene Aliases
2310066E11Rik
Chromosome Location
Chr.6: 83057731 - 83067219 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033606.3 NP_291084.1 2 - 3 601 84
GenBank mRNA AF318278.1 - 1 - 2 245 84
AK010062.1 - 2 - 3 293 84
BC075680.1 - 2 - 3 601 84

View Full Product Details