Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stambp
Entrez Gene ID
70527
Gene Name
STAM binding protein
Gene Aliases
5330424L14Rik, 5730422L11Rik, AW107289, Amsh, mKIAA4198
Chromosome Location
Chr.6: 83543206 - 83575770 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024239.2 NP_077201.1 7 - 8 1105 60
GenBank mRNA AB010123.1 - 7 - 8 1052 60
AK017600.1 - 7 - 8 1060 60
AK019907.1 - 7 - 8 1055 60
AK046995.1 - 7 - 8 1056 60
AK136961.1 - 7 - 8 1656 60
AK139391.1 - 7 - 8 1080 60
AK168810.1 - 7 - 8 1078 60
AK220541.1 - 5 - 6 7336 60
BC003497.1 - 7 - 8 1048 60
BC006939.1 - 7 - 8 1103 60
BC025111.1 - 7 - 8 1047 60

View Full Product Details