The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025748.4 | NP_080024.3 | 5 - 6 | 893 | 131 |
GenBank mRNA | AK016938.1 | - | 5 - 6 | 908 | 131 |
AK021291.1 | - | 5 - 6 | 581 | 131 | |
BC062195.1 | - | 5 - 6 | 611 | 131 |