Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdik1l
Entrez Gene ID
230809
Gene Name
PDLIM1 interacting kinase 1 like
Gene Aliases
BC027088, stk35l2
Chromosome Location
Chr.4: 134275002 - 134287846 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163794.1 NP_001157266.1 2 - 3 399 78
NM_146156.3 NP_666268.1 2 - 3 526 78
GenBank mRNA AK139413.1 - 2 - 3 396 78
AK141566.1 - 2 - 3 389 78
AK141823.1 - 2 - 3 400 78
AK157013.1 - 2 - 3 372 78
AK164975.1 - 2 - 3 578 78
BC027088.1 - 2 - 3 373 78
BC058514.1 - 2 - 3 512 78

View Full Product Details