The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144527.3 | NP_653110.3 | 12 - 13 | 2059 | 64 |
GenBank mRNA | AK030742.1 | - | 12 - 13 | 2024 | 64 |
AK141103.1 | - | 12 - 13 | 2049 | 64 | |
BC021483.1 | - | 1 - 2 | 121 | 64 | |
BC031729.1 | - | 12 - 13 | 1949 | 64 |