Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep85
Entrez Gene ID
70012
Gene Name
centrosomal protein 85
Gene Aliases
2410030J07Rik, AI173272, AI173504, Ccdc21
Chromosome Location
Chr.4: 134129858 - 134187085 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144527.3 NP_653110.3 3 - 4 366 61
GenBank mRNA AK085545.1 - 3 - 4 313 61
AK141103.1 - 3 - 4 356 61
BC008221.1 - 3 - 4 318 61
BC031729.1 - 3 - 4 262 61

View Full Product Details