Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep85
Entrez Gene ID
70012
Gene Name
centrosomal protein 85
Gene Aliases
2410030J07Rik, AI173272, AI173504, Ccdc21
Chromosome Location
Chr.4: 134129858 - 134187085 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144527.3 NP_653110.3 9 - 10 1801 78
GenBank mRNA AK030742.1 - 9 - 10 1766 78
AK141103.1 - 9 - 10 1791 78
BC031729.1 - 9 - 10 1691 78
BC114391.1 - 5 - 6 697 78

View Full Product Details