Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stc2
Entrez Gene ID
20856
Gene Name
stanniocalcin 2
Gene Aliases
AW125853, Stc2l, mustc2
Chromosome Location
Chr.11: 31359441 - 31370061 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011491.3 NP_035621.1 1 - 2 357 75
GenBank mRNA AF031035.1 - 1 - 2 357 75
AF056244.1 - 1 - 2 204 75
AK002527.1 - 1 - 2 165 75
AK010318.1 - 1 - 2 172 75
BC012206.1 - 1 - 2 189 75

View Full Product Details