Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Armc2
Entrez Gene ID
213402
Gene Name
armadillo repeat containing 2
Gene Aliases
2610018I05Rik
Chromosome Location
Chr.10: 41907755 - 42018382 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034858.3 NP_001030030.2 17 - 18 2639 84
GenBank mRNA AK131686.1 - 13 - 14 1837 84
AK141039.1 - 11 - 12 2971 84
BC172168.1 - 12 - 13 2381 84

View Full Product Details