Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lace1
Entrez Gene ID
215951
Gene Name
lactation elevated 1
Gene Aliases
CG8520
Chromosome Location
Chr.10: 42312581 - 42478584 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145743.2 NP_665686.2 12 - 13 1480 99
GenBank mRNA AF397909.1 - 11 - 12 1183 99
AF520417.1 - 12 - 13 1329 99
AK044595.1 - 12 - 13 1356 99
AK170996.1 - 13 - 14 1523 99
BC023308.1 - 11 - 12 1157 99
BC089595.1 - 12 - 13 1330 99

View Full Product Details