Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Qrsl1
Entrez Gene ID
76563
Gene Name
glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1
Gene Aliases
2700038P16Rik, C80053, GatA
Chromosome Location
Chr.10: 43874188 - 43901739 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081054.2 NP_001074523.1 1 - 2 125 50
GenBank mRNA AK012351.1 - 1 - 2 126 50
AK038801.1 - 1 - 2 134 50
AK041347.1 - 1 - 2 130 50
AK170225.1 - 1 - 2 70 50
BC070459.1 - 1 - 2 116 50

View Full Product Details