The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081054.2 | NP_001074523.1 | 4 - 5 | 475 | 83 |
GenBank mRNA | AK012351.1 | - | 4 - 5 | 476 | 83 |
AK038801.1 | - | 4 - 5 | 484 | 83 | |
AK041347.1 | - | 4 - 5 | 480 | 83 | |
AK170225.1 | - | 4 - 5 | 420 | 83 | |
BC070459.1 | - | 4 - 5 | 466 | 83 |