Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snrnp40
Entrez Gene ID
66585
Gene Name
small nuclear ribonucleoprotein 40 (U5)
Gene Aliases
0610009C03Rik, Prp8bp, Wdr57
Chromosome Location
Chr.4: 130360132 - 130390033 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025645.2 NP_079921.2 4 - 5 606 117
GenBank mRNA AK004277.1 - 4 - 5 536 117
AK136135.1 - 4 - 5 536 117
AK152287.1 - 4 - 5 570 117
AK152352.1 - 4 - 5 564 117
AK169605.1 - 4 - 5 552 117
AK207736.1 - 1 - 2 133 117
BC058365.1 - 4 - 5 538 117

View Full Product Details