The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253386.1 | NP_001240315.1 | 4 - 5 | 568 | 63 |
NM_172702.3 | NP_766290.2 | 4 - 5 | 563 | 63 | |
GenBank mRNA | AK075863.1 | - | 4 - 5 | 531 | 63 |
BC031720.1 | - | 4 - 5 | 544 | 63 | |
BG077215.2 | - | 4 - 5 | 518 | 63 | |
BU511121.1 | - | 4 - 5 | 499 | 63 |