The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027928.1 | NP_082204.1 | 3 - 3 | 962 | 78 |
GenBank mRNA | AK004401.1 | - | 1 - 1 | 244 | 78 |