Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clec2e
Entrez Gene ID
232409
Gene Name
C-type lectin domain family 2, member e
Gene Aliases
Clra, clr-a
Chromosome Location
Chr.6: 129091816 - 129100903 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153506.4 NP_705726.3 3 - 4 426 129
GenBank mRNA BC051091.1 - 3 - 4 440 129

View Full Product Details