Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- E230015B07Rik
- Entrez Gene ID
-
320001
- Gene Name
- RIKEN cDNA E230015B07 gene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 87672370 - 87679920 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK042791.1 |
- |
4 - 5 |
409 |
61 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BB639992.1 |
- |
4 - 5 |
409 |
|
Chr. 6 - 87672382 - 87679135 [+] on Build GRCm38 |
61 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
Riken cDNA 1810020O05 gene |
1810020O05Rik |
|
|
Anti-sense |
Chr. - 87675593 - 87690847 on Build GRCm38 |
Downstream |
EF hand and coiled-coil domain containing 1 |
Efcc1 |
Ccdc48 |
54.5kb
|
Sense |
Chr. - 87730820 - 87755912 on Build GRCm38 |
View Full Product Details