The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080469.4 | NP_536717.2 | 2 - 3 | 443 | 65 |
GenBank mRNA | AK083017.1 | - | 2 - 3 | 314 | 65 |
AK144107.1 | - | 2 - 3 | 445 | 65 | |
BC096545.1 | - | 2 - 3 | 2590 | 65 | |
BC116685.1 | - | 2 - 3 | 290 | 65 |