Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ipo13
Entrez Gene ID
230673
Gene Name
importin 13
Gene Aliases
Imp13, Kap13, Ranbp13
Chromosome Location
Chr.4: 117894486 - 117915176 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146152.3 NP_666264.1 5 - 6 1904 109
GenBank mRNA AK052257.1 - 5 - 6 1905 109
AK129200.1 - 5 - 6 1866 109
AK149978.1 - 5 - 6 1887 109
BC031823.1 - 5 - 6 1303 109

View Full Product Details