Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ipo13
Entrez Gene ID
230673
Gene Name
importin 13
Gene Aliases
Imp13, Kap13, Ranbp13
Chromosome Location
Chr.4: 117894486 - 117915176 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146152.3 NP_666264.1 19 - 20 3427 74
GenBank mRNA AK052257.1 - 19 - 20 3428 74
AK129200.1 - 18 - 19 3675 74
AK149978.1 - 19 - 20 3410 74
AK189277.1 - 4 - 5 313 74
AK192686.1 - 4 - 5 393 74
BC031823.1 - 19 - 20 2826 74

View Full Product Details