The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011156.2 | NP_035286.1 | 13 - 14 | 1803 | 53 |
GenBank mRNA | AB007631.1 | - | 13 - 14 | 1686 | 53 |
AK053816.1 | - | 13 - 14 | 1814 | 53 | |
AK076257.1 | - | 13 - 14 | 1802 | 53 | |
AK133875.1 | - | 13 - 14 | 1803 | 53 | |
AK141851.1 | - | 13 - 14 | 1812 | 53 | |
AK161528.1 | - | 13 - 14 | 1802 | 53 | |
AK163774.1 | - | 5 - 6 | 679 | 53 | |
AK163787.1 | - | 13 - 14 | 1815 | 53 | |
AK170570.1 | - | 13 - 14 | 1812 | 53 | |
AK185116.1 | - | 1 - 2 | 110 | 53 | |
AK187379.1 | - | 2 - 3 | 231 | 53 | |
BC012869.1 | - | 13 - 14 | 1803 | 53 | |
BC050830.1 | - | 13 - 14 | 1796 | 53 |