Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dusp9
Entrez Gene ID
75590
Gene Name
dual specificity phosphatase 9
Gene Aliases
Dusp4, Mpk4, Pyst3
Chromosome Location
Chr.X: 73639389 - 73643514 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029352.3 NP_083628.3 2 - 3 814 58
GenBank mRNA AK141667.1 - 2 - 3 839 58
AY316312.1 - 2 - 3 814 58
BC100309.1 - 2 - 3 814 58

View Full Product Details