Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sesn2
Entrez Gene ID
230784
Gene Name
sestrin 2
Gene Aliases
HI95, SEST2, Ses2
Chromosome Location
Chr.4: 132492804 - 132510456 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144907.1 NP_659156.1 8 - 9 1506 120
GenBank mRNA AK143788.1 - 8 - 9 1550 120
AK170228.1 - 8 - 9 1519 120
AK170547.1 - - 1550 120
AK184702.1 - 1 - 2 120
BC005672.1 - 8 - 9 1506 120

View Full Product Details