The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166547.1 | NP_001160019.1 | 18 - 19 | 2943 | 56 |
NM_001166548.1 | NP_001160020.1 | 17 - 18 | 2875 | 56 | |
NM_001166549.1 | NP_001160021.1 | 17 - 18 | 2979 | 56 | |
NM_023743.2 | NP_076232.2 | 18 - 19 | 2958 | 56 | |
GenBank mRNA | AB031388.1 | - | 18 - 19 | 2779 | 56 |
AK012082.1 | - | 8 - 9 | 1362 | 56 | |
AK041598.1 | - | 17 - 18 | 2837 | 56 | |
AK088425.1 | - | 13 - 14 | 2045 | 56 | |
AK128990.1 | - | 17 - 18 | 2975 | 56 | |
AK165678.1 | - | 18 - 19 | 2945 | 56 | |
BC033410.1 | - | 18 - 19 | 2793 | 56 |