The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253690.1 | NP_001240619.1 | 4 - 5 | 1738 | 69 |
NR_045108.2 | 5 - 6 | 619 | 69 | ||
GenBank mRNA | BC055947.1 | - | 5 - 6 | 469 | 69 |