The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130458.1 | NP_001123930.1 | 2 - 3 | 260 | 82 |
NM_001130459.1 | NP_001123931.1 | 2 - 3 | 229 | 82 | |
NM_015749.3 | NP_056564.1 | 2 - 3 | 263 | 82 | |
GenBank mRNA | AF090686.1 | - | 1 - 2 | 190 | 82 |
AK133707.1 | - | 2 - 3 | 225 | 82 | |
AK147176.1 | - | 2 - 3 | 234 | 82 | |
AK161618.1 | - | 2 - 3 | 230 | 82 | |
AK170401.1 | - | 2 - 3 | 249 | 82 | |
AK197212.1 | - | 1 - 2 | 183 | 82 | |
BB849607.1 | - | 2 - 3 | 258 | 82 | |
BC003720.1 | - | 1 - 2 | 136 | 82 |