The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172438.3 | NP_766026.1 | 19 - 20 | 2049 | 141 |
GenBank mRNA | AK050734.1 | - | 19 - 20 | 2049 | 141 |
AK173078.1 | - | 17 - 18 | 1887 | 141 | |
BC020332.1 | - | 18 - 19 | 2014 | 141 | |
BC021779.1 | - | 18 - 19 | 2022 | 141 | |
BC039758.1 | - | 19 - 20 | 2050 | 141 |