Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Smek2
- Entrez Gene ID
-
104570
- Gene Name
- SMEK homolog 2, suppressor of mek1 (Dictyostelium)
- Gene Aliases
-
AW011752, AW557776, mKIAA1387
- Chromosome Location
-
Chr.11: 29167939 - 29220797 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK132660.1 |
- |
2 - 3 |
119 |
149 |
View Full Product Details