The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029861.2 | NP_084137.1 | 1 - 2 | 413 | 63 |
GenBank mRNA | AK005381.1 | - | 2 - 3 | 477 | 63 |
AK148349.1 | - | 1 - 2 | 489 | 63 | |
AK171947.1 | - | 1 - 2 | 463 | 63 | |
AK204024.1 | - | 1 - 2 | 52 | 63 | |
BC087946.1 | - | 1 - 2 | 414 | 63 |