The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282993.1 | NP_001269922.1 | 2 - 3 | 483 | 64 |
NM_001282994.1 | NP_001269923.1 | 2 - 3 | 314 | 64 | |
NM_172496.3 | NP_766084.3 | 2 - 3 | 483 | 64 | |
GenBank mRNA | AK028833.1 | - | 2 - 3 | 330 | 64 |
AK049730.1 | - | 2 - 3 | 276 | 64 | |
AK122334.1 | - | 2 - 3 | 314 | 64 | |
AK147366.1 | - | 2 - 3 | 484 | 64 | |
AK147633.1 | - | 2 - 3 | 484 | 64 | |
AY308745.1 | - | 2 - 3 | 323 | 64 | |
BC145116.1 | - | 2 - 3 | 323 | 64 | |
BC145118.1 | - | 2 - 3 | 323 | 64 | |
BC150706.1 | - | 2 - 3 | 323 | 64 | |
U26967.1 | - | 1 - 2 | 56 | 64 |