The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194053.3 | NP_918942.1 | 1 - 2 | 799 | 85 |
NM_194054.3 | NP_918943.1 | 1 - 2 | 799 | 85 | |
GenBank mRNA | AK220511.1 | - | 1 - 2 | 759 | 85 |
AY102282.1 | - | 1 - 2 | 756 | 85 | |
AY102284.1 | - | 1 - 2 | 756 | 85 | |
AY114152.1 | - | 1 - 2 | 754 | 85 | |
BC056373.1 | - | 1 - 2 | 627 | 85 |