The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162432.1 | NP_001155904.1 | 2 - 3 | 194 | 78 |
NM_001162433.1 | NP_001155905.1 | 2 - 3 | 236 | 78 | |
NM_010693.3 | NP_034823.1 | 1 - 2 | 291 | 78 | |
GenBank mRNA | AK088001.1 | - | 1 - 2 | 292 | 78 |
AK134013.1 | - | 1 - 2 | 281 | 78 | |
BC011474.1 | - | 2 - 3 | 201 | 78 | |
BQ563042.1 | - | 2 - 3 | 185 | 78 | |
BY087238.1 | - | 2 - 3 | 195 | 78 | |
BY100584.1 | - | 2 - 3 | 238 | 78 | |
M12056.1 | - | 1 - 2 | 267 | 78 | |
X03533.1 | - | 1 - 2 | 297 | 78 |