Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf19b
Entrez Gene ID
75234
Gene Name
ring finger protein 19B
Gene Aliases
4930534K13Rik, 4930555L03Rik, Ibrdc3
Chromosome Location
Chr.4: 129058174 - 129085886 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029219.1 NP_083495.1 3 - 4 979 92
GenBank mRNA BC117807.1 - 3 - 4 537 92
BC139808.1 - 3 - 4 540 92


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC038660.1 - Chr. - - [-] on Build 92

View Full Product Details