Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cited2
- Entrez Gene ID
-
17684
- Gene Name
- Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2
- Gene Aliases
-
AI835299, ER154-like, Mrg1, Msg2, p35srj
- Chromosome Location
-
Chr.10: 17723205 - 17725674 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U86445.1 |
- |
2 - 3 |
695 |
66 |
View Full Product Details