The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031185.3 | NP_112462.1 | 3 - 4 | 5246 | 95 |
GenBank mRNA | AB020886.1 | - | 3 - 4 | 5195 | 95 |
AB025278.1 | - | 3 - 4 | 5209 | 95 | |
AB051563.1 | - | 3 - 4 | 5197 | 95 | |
AB070852.1 | - | 2 - 3 | 4931 | 95 | |
AB070853.1 | - | 2 - 3 | 4917 | 95 | |
AF326228.1 | - | 3 - 4 | 5195 | 95 | |
AF326230.1 | - | 2 - 3 | 4900 | 95 | |
BC042461.1 | - | 1 - 2 | 4642 | 95 | |
BC043939.1 | - | 1 - 2 | 4642 | 95 | |
BC071185.1 | - | 3 - 4 | 5194 | 95 | |
BC138364.1 | - | 3 - 4 | 5191 | 95 | |
BC138365.1 | - | 3 - 4 | 5190 | 95 |