Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sepn1
Entrez Gene ID
74777
Gene Name
selenoprotein N, 1
Gene Aliases
1110019I12Rik, AI414492
Chromosome Location
Chr.4: 134537892 - 134552166 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029100.2 NP_083376.2 7 - 8 1038 95
GenBank mRNA CF737950.1 - 1 - 2 80 95
CX562316.1 - 6 - 7 715 95

View Full Product Details